PRIH93ED82CA Princeton BioMolecular Research, Inc.

3-(2,5-dioxoimidazolidin-4-yl)cyclopentanecarboxamide

  • MDL number MFCD31944915
  • Molecular Weight : 211.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart