PRIH93ED82DF Princeton BioMolecular Research, Inc.

5,6-dichloro-2-ethyl-1H-benzimidazole

  • MDL number MFCD05992934
  • Molecular Weight : 215.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart