PRIH93ED82F0 Princeton BioMolecular Research, Inc.

2-(4-fluorophenoxy)-1-(2,4,6-trihydroxyphenyl)ethanone

  • MDL number MFCD00422145
  • Molecular Weight : 278.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart