PRIH93ED82F8 Princeton BioMolecular Research, Inc.

3-(5-methyl-2-furyl)-1H-pyrazole

  • MDL number MFCD06808814
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart