PRIH93ED8310 Princeton BioMolecular Research, Inc.

3-(3,5-dimethyl-1H-pyrazol-1-yl)-4-methoxybenzoic acid

  • MDL number MFCD21998805
  • Molecular Weight : 246.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart