PRIH93ED8314 Princeton BioMolecular Research, Inc.

5-amino-4-(1H-benzimidazol-2-yl)-1-(2-furylmethyl)-1,2-dihydro-3H-pyrrol-3-one

  • MDL number MFCD02704941
  • Molecular Weight : 294.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart