PRIH93ED8318 Princeton BioMolecular Research, Inc.

(8-fluoro-1,3,4,5-tetrahydro-2H-pyrido[4,3-b]indol-2-yl)(oxo)acetic acid

  • MDL number MFCD11982952
  • Molecular Weight : 262.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart