PRIH93ED831F Princeton BioMolecular Research, Inc.

(1R,5S)-decahydro-8H-1,5-methanopyrido[1,2-a][1,5]diazocin-8-one

  • MDL number MFCD31806893
  • Molecular Weight : 194.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart