PRIH93ED8326 Princeton BioMolecular Research, Inc.

2-[(1H-benzimidazol-2-ylmethyl)thio]-6-propylpyrimidin-4(1H)-one

  • MDL number MFCD01788335
  • Molecular Weight : 300.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart