PRIH93ED8333 Princeton BioMolecular Research, Inc.

3-({1-[2-(4-methylphenoxy)ethyl]-1H-benzimidazol-2-yl}thio)propanoic acid

  • MDL number MFCD01072845
  • Molecular Weight : 356.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart