PRIH93ED8334 Princeton BioMolecular Research, Inc.

1-[2-(3-methylphenoxy)ethyl]-1H-benzimidazole-2-sulfonic acid

  • MDL number MFCD03828497
  • Molecular Weight : 332.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart