PRIH93ED833B Princeton BioMolecular Research, Inc.

2-(2-{[2-(4-ethoxyphenoxy)ethyl]thio}-1H-benzimidazol-1-yl)acetamide

  • MDL number MFCD02646685
  • Molecular Weight : 371.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart