PRIH93ED8340 Princeton BioMolecular Research, Inc.

1-[2-(4-ethoxyphenoxy)ethyl]-1H-benzimidazole-2-thiol

  • MDL number MFCD03724201
  • Molecular Weight : 314.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart