PRIH93ED8345 Princeton BioMolecular Research, Inc.

({6-amino-9-[2-(4-ethoxyphenoxy)ethyl]-9H-purin-8-yl}thio)acetic acid

  • MDL number MFCD03765031
  • Molecular Weight : 389.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart