PRIH93ED834C Princeton BioMolecular Research, Inc.

2-({6-amino-9-[2-(4-methylphenoxy)ethyl]-9H-purin-8-yl}thio)ethanol

  • MDL number MFCD02908146
  • Molecular Weight : 345.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart