PRIH93ED834D Princeton BioMolecular Research, Inc.

8-(benzylthio)-9-[2-(4-isopropoxyphenoxy)ethyl]-9H-purin-6-amine

  • MDL number MFCD06245076
  • Molecular Weight : 435.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart