PRIH93ED834E Princeton BioMolecular Research, Inc.

8-{[2-(2-methylphenoxy)ethyl]thio}-9H-purin-6-amine

  • MDL number MFCD00621744
  • Molecular Weight : 301.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart