PRIH93ED8380 Princeton BioMolecular Research, Inc.

2-{2-[(2-methyl-1H-indol-3-yl)methyl]-1H-benzimidazol-1-yl}ethanol

  • MDL number MFCD30594625
  • Molecular Weight : 305.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart