PRIH93ED8395 Princeton BioMolecular Research, Inc.

4-{[(3,5-dimethylisoxazol-4-yl)sulfonyl]amino}benzoic acid

  • MDL number MFCD13563196
  • Molecular Weight : 296.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart