PRIH93ED8398 Princeton BioMolecular Research, Inc.

1-(3-iodobenzoyl)proline

  • MDL number MFCD07748945
  • Molecular Weight : 345.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart