PRIH93ED83BC Princeton BioMolecular Research, Inc.

(2E)-2-{[(E)-amino(imino)methyl]hydrazono}propanoic acid

  • MDL number MFCD01361002
  • Molecular Weight : 144.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart