PRIH93ED83C9 Princeton BioMolecular Research, Inc.

[(1H-indol-3-ylmethyl)thio]acetic acid

  • MDL number MFCD03177204
  • Molecular Weight : 221.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart