PRIH93ED83D1 Princeton BioMolecular Research, Inc.

3'-(4-bromophenyl)-5'-methyl-4'H-spiro[indole-3,2'-[1,3]thiazolidine]-2,4'(1H)-dione

  • MDL number MFCD00834615
  • Molecular Weight : 389.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart