PRIH93ED83DD Princeton BioMolecular Research, Inc.

((2E)-2-{(2Z)-[1-(2-furyl)ethylidene]hydrazono}-4-oxo-1,3-thiazolidin-5-yl)acetic acid

  • MDL number MFCD31808858
  • Molecular Weight : 281.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart