PRIH93ED83E5 Princeton BioMolecular Research, Inc.

dispiro[bicyclo[2.2.1]hept-5-ene-2,5'-[1,3]dioxane-2',3''-indol]-2''(1''H)-one

  • MDL number MFCD02103122
  • Molecular Weight : 283.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart