PRIH93ED83FE Princeton BioMolecular Research, Inc.

3,3'-[(4-fluorophenyl)methylene]bis(2-methyl-1H-indole)

  • MDL number MFCD01464673
  • Molecular Weight : 368.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart