PRIH93ED8407 Princeton BioMolecular Research, Inc.

2-{[(1E)-1,3-benzodioxol-5-ylmethylene]amino}-5-bromobenzoic acid

  • MDL number MFCD00755097
  • Molecular Weight : 348.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart