PRIH93ED8417 Princeton BioMolecular Research, Inc.

3,3'-dithiobis(4-nitrobenzoic acid)

  • MDL number MFCD00517926
  • Molecular Weight : 396.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart