PRIH93ED841C Princeton BioMolecular Research, Inc.

dimethyl 4,4'-dithiobis(3-nitrobenzoate)

  • MDL number MFCD00025862
  • Molecular Weight : 424.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart