PRIH93ED8434 Princeton BioMolecular Research, Inc.

methyl 5-nitro-2-(pyridin-2-ylthio)benzoate

  • MDL number MFCD06203744
  • Molecular Weight : 290.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart