PRIH93ED8436 Princeton BioMolecular Research, Inc.

methyl 4-[(4-methoxyphenyl)amino]-3-nitrobenzoate

  • MDL number MFCD30707347
  • Molecular Weight : 302.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart