PRIH93ED843B Princeton BioMolecular Research, Inc.

2-[(2-anilino-2-oxoethyl)thio]-N-[4-(pentyloxy)phenyl]acetamide

  • MDL number MFCD00363001
  • Molecular Weight : 386.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart