PRIH93ED8444 Princeton BioMolecular Research, Inc.

methyl 2-[({[5-(3-fluorophenyl)-4-phenyl-4H-1,2,4-triazol-3-yl]thio}acetyl)amino]benzoate

  • MDL number MFCD00323518
  • Molecular Weight : 462.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart