PRIH93ED8447 Princeton BioMolecular Research, Inc.

N-(4-fluorophenyl)-2-{[5-(2-furyl)-4-phenyl-4H-1,2,4-triazol-3-yl]thio}acetamide

  • MDL number MFCD00350021
  • Molecular Weight : 394.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart