PRIH93ED844D Princeton BioMolecular Research, Inc.

3-(2-chlorophenyl)-2-[(pyridin-2-ylthio)methyl]quinazolin-4(3H)-one

  • MDL number MFCD00372270
  • Molecular Weight : 379.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart