PRIH93ED8450 Princeton BioMolecular Research, Inc.

2-[(6-ethoxy-1,3-benzothiazol-2-yl)thio]-N-(4-ethylphenyl)acetamide

  • MDL number MFCD00741782
  • Molecular Weight : 372.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart