PRIH93ED845C Princeton BioMolecular Research, Inc.

2-{[5-(3,4-dimethoxyphenyl)-4-phenyl-4H-1,2,4-triazol-3-yl]thio}-N-(2,3-dimethylphenyl)acetamide

  • MDL number MFCD00754619
  • Molecular Weight : 474.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart