PRIH93ED8465 Princeton BioMolecular Research, Inc.

N-(5-isopropyl-4-methoxy-2-methylphenyl)-3-methoxybenzamide

  • MDL number MFCD01429296
  • Molecular Weight : 313.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart