PRIH93ED8472 Princeton BioMolecular Research, Inc.

5-[(4-nitrobenzyl)thio]-1-phenyl-1H-tetrazole

  • MDL number MFCD00991851
  • Molecular Weight : 313.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart