PRIH93ED8475 Princeton BioMolecular Research, Inc.

2-[(2-anilino-2-oxoethyl)thio]-N-(2,4-dimethylphenyl)acetamide

  • MDL number MFCD02106966
  • Molecular Weight : 328.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart