PRIH93ED847A Princeton BioMolecular Research, Inc.

2-cyclobutyl-N-(4-ethoxyphenyl)acetamide

  • MDL number MFCD00442949
  • Molecular Weight : 233.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart