PRIH93ED8491 Princeton BioMolecular Research, Inc.

N-[2-(3-methoxyphenoxy)ethyl]octadecanamide

  • MDL number MFCD00333797
  • Molecular Weight : 433.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart