PRIH93ED8498 Princeton BioMolecular Research, Inc.

4-bromo-N-[4-(pentyloxy)phenyl]benzamide

  • MDL number MFCD00402499
  • Molecular Weight : 362.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart