PRIH93ED849C Princeton BioMolecular Research, Inc.

4-butoxy-N-(2-isopropoxyphenyl)benzamide

  • MDL number MFCD00382995
  • Molecular Weight : 327.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart