PRIH93ED84A2 Princeton BioMolecular Research, Inc.

N-(4-bromo-2,6-dimethylphenyl)-3,4,5-trimethoxybenzamide

  • MDL number MFCD00383015
  • Molecular Weight : 394.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart