PRIH93ED84A6 Princeton BioMolecular Research, Inc.

2-(4-ethylphenoxy)-N-(1-naphthylmethyl)acetamide

  • MDL number MFCD00383038
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart