PRIH93ED84AF Princeton BioMolecular Research, Inc.

2,4-dichloro-N-[4-(pyridin-4-ylmethyl)phenyl]benzamide

  • MDL number MFCD00741779
  • Molecular Weight : 357.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart