PRIH93ED84B2 Princeton BioMolecular Research, Inc.

N-(1,5-dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)-1-naphthamide

  • MDL number MFCD00362917
  • Molecular Weight : 357.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart