PRIH93ED84B5 Princeton BioMolecular Research, Inc.

4-butoxy-N-(4-isopropylbenzyl)benzamide

  • MDL number MFCD00384577
  • Molecular Weight : 325.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart