PRIH93ED84C0 Princeton BioMolecular Research, Inc.

4-butoxy-N-(2-furylmethyl)benzamide

  • MDL number MFCD00423420
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart